missing translation for 'onlineSavingsMsg'
Learn More

3-Bromobenzyl mercaptan, 98%, Thermo Scientific™

Product Code. 11431270
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Solubility
Insoluble in water. Soluble in toluene.

Notes
Air Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, air.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 886497-84-3
Molecular Formula C7H7BrS
Molecular Weight (g/mol) 203.097
MDL Number MFCD06798010
InChI Key LXEHTZZVGCUFCS-UHFFFAOYSA-N
Synonym 3-bromophenyl methanethiol, 3-bromobenzylmercaptan, 3-bromobenzyl mercaptan, 3-bromo-alpha-toluenethiol, benzenemethanethiol,3-bromo, 3-bromophenyl methane-1-thiol
PubChem CID 2761417
IUPAC Name (3-bromophenyl)methanethiol
SMILES C1=CC(=CC(=C1)Br)CS

Specifications

Quantity 1 g
UN Number UN2810
Sensitivity Air Sensitive
Solubility Information Insoluble in water. Soluble in toluene.
Formula Weight 203.1
Percent Purity 98%
Chemical Name or Material 3-Bromobenzyl mercaptan
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'