missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Bromoimidazo[1,2-a]pyridine-6-carboxylic acid hydrate, 95%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
Dimensione della confezione:
1g
250mg
Descrizione
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifica
Specifica
| Melting Point | 297°C to 299°C |
| Quantity | 250 mg |
| Formula Weight | 241.05 (Anhydrous) |
| Percent Purity | 95% |
| Chemical Name or Material | 3-Bromoimidazo[1,2-a]pyridine-6-carboxylic acid hydrate |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Spot an opportunity for improvement?Share a Content Correction