missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-4-(trifluoromethoxy)phenylacetic acid, 97%, Thermo Scientific™

Product Code. 15488085
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 15488085

Brand: Thermo Scientific Alfa Aesar H32437.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1017779-77-9
Molecular Formula C9H6ClF3O3
Molecular Weight (g/mol) 254.59
MDL Number MFCD09832392
InChI Key MYYLIFIHZIRCPZ-UHFFFAOYSA-N
Synonym 3-chloro-4-trifluoromethoxy phenylacetic acid, 2-3-chloro-4-trifluoromethoxy phenyl acetic acid, 3-chloro-4-trifluoromethoxy phenyl acetic acid, 3-chloro-4-trifluoromethoxy phenylacetic acid, jrd
PubChem CID 20112199
IUPAC Name 2-[3-chloro-4-(trifluoromethoxy)phenyl]acetic acid
SMILES OC(=O)CC1=CC(Cl)=C(OC(F)(F)F)C=C1

Specifications

Melting Point 72°C to 76°C
Quantity 1 g
Formula Weight 254.59
Percent Purity 97%
Chemical Name or Material 3-Chloro-4-(trifluoromethoxy)phenylacetic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.