Learn More
3-Chloro-5-(trifluoromethyl)benzylamine, 97%, Thermo Scientific™
99.76 EUR - 435.00 EUR
Chemical Identifiers
CAS | 400771-41-7 |
---|---|
Molecular Formula | C8H7ClF3N |
Molecular Weight (g/mol) | 209.596 |
MDL Number | MFCD07786683 |
InChI Key | GCLAXYMPBCHQIK-UHFFFAOYSA-N |
Synonym | 3-chloro-5-trifluoromethyl phenyl methanamine, 3-chloro-5-trifluoromethyl-benzylamine, 1-3-chloro-5-trifluoromethyl phenyl methanamine, 3-chloro-5-trifluoromethyl benzylamine, 3-chloro-5-trifluoromethyl benzenemethanamine |
PubChem CID | 18614348 |
IUPAC Name | [3-chloro-5-(trifluoromethyl)phenyl]methanamine |
SMILES | C1=C(C=C(C=C1C(F)(F)F)Cl)CN |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15477367
|
Thermo Scientific Alfa Aesar
H34423.03 |
1 g | N/A | ||||||
15487367
|
Thermo Scientific Alfa Aesar
H34423.06 |
5 g |
435.00 EUR
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications3-Chloro-5-(trifluoromethyl)benzylamine is used as an organic chemical synthesis intermediate.
Notes
Air sensitive. Incompatible with oxidizing agents and acids.
Chemical Identifiers
400771-41-7 | |
209.596 | |
GCLAXYMPBCHQIK-UHFFFAOYSA-N | |
18614348 | |
C1=C(C=C(C=C1C(F)(F)F)Cl)CN |
C8H7ClF3N | |
MFCD07786683 | |
3-chloro-5-trifluoromethyl phenyl methanamine, 3-chloro-5-trifluoromethyl-benzylamine, 1-3-chloro-5-trifluoromethyl phenyl methanamine, 3-chloro-5-trifluoromethyl benzylamine, 3-chloro-5-trifluoromethyl benzenemethanamine | |
[3-chloro-5-(trifluoromethyl)phenyl]methanamine |
Specifications
400771-41-7 | |
MFCD07786683 | |
Air sensitive | |
GCLAXYMPBCHQIK-UHFFFAOYSA-N | |
[3-chloro-5-(trifluoromethyl)phenyl]methanamine | |
18614348 | |
97% |
C8H7ClF3N | |
UN2735 | |
3-chloro-5-trifluoromethyl phenyl methanamine, 3-chloro-5-trifluoromethyl-benzylamine, 1-3-chloro-5-trifluoromethyl phenyl methanamine, 3-chloro-5-trifluoromethyl benzylamine, 3-chloro-5-trifluoromethyl benzenemethanamine | |
C1=C(C=C(C=C1C(F)(F)F)Cl)CN | |
209.596 | |
209.6 | |
3-Chloro-5-(trifluoromethyl)benzylamine |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures; Store under Nitrogen