missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-6-methoxybenzo[b]thiophene-2-carbonyl chloride, Thermo Scientific™

72.13 EUR - 256.00 EUR

Chemical Identifiers

CAS 75998-29-7
Molecular Formula C10H6Cl2O2S
Molecular Weight (g/mol) 261.116
MDL Number MFCD00448907
InChI Key DTMXTCKEPQWRPO-UHFFFAOYSA-N
Synonym 3-chloro-6-methoxybenzo b thiophene-2-carbonyl chloride, benzo b thiophene-2-carbonyl chloride, 3-chloro-6-methoxy, acmc-20an6b, aronis24536, 3-chloro-6-methoxy-benzo b thiophene-2-carbonyl chloride, 3-chloro-6-methoxybenzo-b thiophene-2-carbonyl chloride, 6-methoxy-3-chlorobenzo b thiophene-2-carbonyl chloride, 6-methoxy-3-chloro-benzo b-thiophene-2-carbonyl chloride, benzo b thiophene-2-carbonylchloride, 3-chloro-6-methoxy
PubChem CID 12594638
IUPAC Name 3-chloro-6-methoxy-1-benzothiophene-2-carbonyl chloride
SMILES COC1=CC2=C(C=C1)C(=C(S2)C(=O)Cl)Cl
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11315439
View Documents Promotion Details
Thermo Scientific Alfa Aesar
H50419.MD
250 mg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
11325439
View Documents Promotion Details
Thermo Scientific Alfa Aesar
H50419.03
1 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

75998-29-7
261.116
DTMXTCKEPQWRPO-UHFFFAOYSA-N
12594638
COC1=CC2=C(C=C1)C(=C(S2)C(=O)Cl)Cl
C10H6Cl2O2S
MFCD00448907
3-chloro-6-methoxybenzo b thiophene-2-carbonyl chloride, benzo b thiophene-2-carbonyl chloride, 3-chloro-6-methoxy, acmc-20an6b, aronis24536, 3-chloro-6-methoxy-benzo b thiophene-2-carbonyl chloride, 3-chloro-6-methoxybenzo-b thiophene-2-carbonyl chloride, 6-methoxy-3-chlorobenzo b thiophene-2-carbonyl chloride, 6-methoxy-3-chloro-benzo b-thiophene-2-carbonyl chloride, benzo b thiophene-2-carbonylchloride, 3-chloro-6-methoxy
3-chloro-6-methoxy-1-benzothiophene-2-carbonyl chloride

Specifications

75998-29-7
MFCD00448907
3-chloro-6-methoxybenzo b thiophene-2-carbonyl chloride, benzo b thiophene-2-carbonyl chloride, 3-chloro-6-methoxy, acmc-20an6b, aronis24536, 3-chloro-6-methoxy-benzo b thiophene-2-carbonyl chloride, 3-chloro-6-methoxybenzo-b thiophene-2-carbonyl chloride, 6-methoxy-3-chlorobenzo b thiophene-2-carbonyl chloride, 6-methoxy-3-chloro-benzo b-thiophene-2-carbonyl chloride, benzo b thiophene-2-carbonylchloride, 3-chloro-6-methoxy
COC1=CC2=C(C=C1)C(=C(S2)C(=O)Cl)Cl
261.116
261.13
C10H6Cl2O2S
UN3261
DTMXTCKEPQWRPO-UHFFFAOYSA-N
3-chloro-6-methoxy-1-benzothiophene-2-carbonyl chloride
12594638
3-Chloro-6-methoxybenzo[b]thiophene-2-carbonyl chloride
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications