missing translation for 'onlineSavingsMsg'
Learn More

3'-Chloropropiophenone, 98%

Product Code. 11464224
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
This item is not returnable. View return policy

Product Code. 11464224

Brand: Thermo Scientific Alfa Aesar A12542.36

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 34841-35-5 | C9H9ClO | 168.62 g/mol

3'-Chloropropiophenone has been used in the preparation of 1-(3-chlorophenyl)-1-phenyl-1-propanol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3′-Chloropropiophenone has been used in the preparation of 1-(3-chlorophenyl)-1-phenyl-1-propanol.

Solubility
Soluble in methanol.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 34841-35-5
Molecular Formula C9H9ClO
Molecular Weight (g/mol) 168.62
MDL Number MFCD00009925
InChI Key PQWGFUFROKIJBO-UHFFFAOYSA-N
Synonym 3'-chloropropiophenone, 1-3-chlorophenyl propan-1-one, m-chloropropiophenone, 3-chlorophenyl ethyl ketone, 1-propanone, 1-3-chlorophenyl, 1-3-chloro-phenyl-propan-1-one, m-chloro-propiophenone, pubchem3083, 3'-chloro-propiophenone, 3/'-chloropropiophenone
PubChem CID 587128
IUPAC Name 1-(3-chlorophenyl)propan-1-one
SMILES CCC(=O)C1=CC(=CC=C1)Cl

Specifications

Melting Point 45°C to 47°C
Boiling Point 123°C to 124°C (13 mmHg)
Flash Point >110°C (230°F)
Quantity 500 g
Solubility Information Soluble in methanol.
Formula Weight 168.62
Percent Purity 98%
Chemical Name or Material 3'-Chloropropiophenone

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.