missing translation for 'onlineSavingsMsg'
Learn More

3-Dimethylamino-1-propanol, 99%, Thermo Scientific™

22.30 EUR - 84.19 EUR

Chemical Identifiers

CAS 3179-63-3
Molecular Formula C5H13NO
Molecular Weight (g/mol) 103.17
MDL Number MFCD00002945
InChI Key PYSGFFTXMUWEOT-UHFFFAOYSA-N
Synonym 3-dimethylamino-1-propanol, 3-dimethylamino propan-1-ol, 1-propanol, 3-dimethylamino, dimethylaminopropanol, n,n-dimethylpropanolamine, 3-dimethylamino propanol, dimethylpropanolamine, 3-dimethlyamino propanol, 3-n,n-dimethylamino propanol
PubChem CID 76646
IUPAC Name 3-(dimethylamino)propan-1-ol
SMILES CN(C)CCCO
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10496910
View Documents
Thermo Scientific Acros
115880050
5 g Glass bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10406920
View Documents
Thermo Scientific Acros
115881000
100 g Glass bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10529420
View Documents
Thermo Scientific Acros
115885000
500 g Glass bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Specifications

Chemical Identifiers

3179-63-3
103.17
PYSGFFTXMUWEOT-UHFFFAOYSA-N
76646
CN(C)CCCO
C5H13NO
MFCD00002945
3-dimethylamino-1-propanol, 3-dimethylamino propan-1-ol, 1-propanol, 3-dimethylamino, dimethylaminopropanol, n,n-dimethylpropanolamine, 3-dimethylamino propanol, dimethylpropanolamine, 3-dimethlyamino propanol, 3-n,n-dimethylamino propanol
3-(dimethylamino)propan-1-ol

Specifications

3179-63-3
100.0
Colorless to Yellow
159.0°C
Authentic
C5H13NO
1.4350 to 1.4370
04, I, 433
3-dimethylamino-1-propanol, 3-dimethylamino propan-1-ol, 1-propanol, 3-dimethylamino, dimethylaminopropanol, n,n-dimethylpropanolamine, 3-dimethylamino propanol, dimethylpropanolamine, 3-dimethlyamino propanol, 3-n,n-dimethylamino propanol
PYSGFFTXMUWEOT-UHFFFAOYSA-N
3-(dimethylamino)propan-1-ol
76646
103.16
Liquid
98.5
-35.0°C
0.8860g/mL
54°C
98.5% min. (GC)
(CH3)2N(CH2)3OH
MFCD00002945
0.886
Solubility in water: soluble
CN(C)CCCO
103.17
6.85 mPa.s (20°C)
99%
3-Dimethylamino-1-propanol, 99%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Flammable liquid and vapor.
Harmful if swallowed.
Causes severe skin burns and eye damage.

GHS P Statement
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 221-659-2

missing translation for 'rtecsNumber' : UB3155000

missing translation for 'tsca' : TSCA

Product Identifier
  • 3-Dimethylamino-1-propanol

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 B

Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
  • H226-Flammable liquid and vapour.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only