missing translation for 'onlineSavingsMsg'
Learn More

3-Ethynylpyridine, 97%

Product Code. 15440308
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 15440308

Brand: Thermo Scientific Alfa Aesar H53465.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2510-23-8 | C7H5N | 103.124 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2510-23-8
Molecular Formula C7H5N
Molecular Weight (g/mol) 103.124
MDL Number MFCD02177459
InChI Key CLRPXACRDTXENY-UHFFFAOYSA-N
Synonym pyridine, 3-ethynyl, 3-pyridylacetylene, 3-ethynyl-pyridine, pyridine, 3-ethynyl-6ci, 7ci, 8ci, 9ci, 3-ethylnylpyridine, 3-ethynyl pyridine, meta-ethynylpyridine, 3-ethylnyl pyridine, 3-pyridyl acetylene
PubChem CID 186003
IUPAC Name 3-ethynylpyridine
SMILES C#CC1=CN=CC=C1

Specifications

Melting Point 39°C to 40°C
Boiling Point 83°C to 84°C (30 mmHg)
Flash Point 57°C (135°F)
Quantity 5 g
UN Number UN1325
Formula Weight 103.12
Percent Purity 97%
Chemical Name or Material 3-Ethynylpyridine
compliance-icons compliance-icons
Product Identifier
  • 3-Ethynylpyridine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H228-Flammable solid.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : 3-Ethynylpyridine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.