missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoro-4-formylphenylboronic acid, 95%, Thermo Scientific™

Product Code. p-4602032 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11956151

Brand: Thermo Scientific Acros 445600010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 248270-25-9
Molecular Formula C7H6BFO3
Molecular Weight (g/mol) 167.93
InChI Key NZNRMUVHUVCIBR-UHFFFAOYSA-N
Synonym 3-fluoro-4-formylbenzeneboronic acid, 3-fluoro-4-formylphenyl boronic acid, 3-fluoro-4-formylphenboronic acid, boronic acid, 3-fluoro-4-formylphenyl, pubchem1847, acmc-1cga0, ksc494k4h, 4-borono-3-fluorobenzaldehyde, 3-fluoro-4-formyl-phenyl boronic acid, 3-fluoranyl-4-methanoyl-phenyl boronic acid
PubChem CID 2778654
IUPAC Name (3-fluoro-4-formylphenyl)boronic acid
SMILES B(C1=CC(=C(C=C1)C=O)F)(O)O

Specifications

Assay Percent Range 94% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 167.93
Percent Purity 95%
Chemical Name or Material 3-Fluoro-4-formylphenylboronic acid
compliance-icons
Product Identifier
  • 3-Fluoro-4-formylphenylboronic acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.