Learn More
3-Fluoro-4-(trifluoromethyl)benzoyl chloride, 97%, Thermo Scientific™
44.80 EUR - 134.00 EUR
Chemical Identifiers
CAS | 216144-68-2 |
---|---|
Molecular Formula | C8H3ClF4O |
Molecular Weight (g/mol) | 226.55 |
MDL Number | MFCD00236284 |
InChI Key | DPDAEMNQJYDHRQ-UHFFFAOYSA-N |
Synonym | 3-fluoro-4-trifluoromethyl benzoyl chloride, 3-fluoro-4-trifluoromethyl benzoylchloride, 3-fluoranyl-4-trifluoromethyl benzoyl chloride, acmc-1cobr, 4-chlorocarbonyl-2-fluorobenzotrifluoride, 3-fluoro-4-trifiuoromethyl benzoyl chloride, 3-fluoro-4-trifluoromethyl benzoyl cloride, benzoylchloride, 3-fluoro-4-trifluoromethyl, benzoyl chloride, 3-fluoro-4-trifluoromethyl |
PubChem CID | 2734709 |
IUPAC Name | 3-fluoro-4-(trifluoromethyl)benzoyl chloride |
SMILES | FC1=CC(=CC=C1C(F)(F)F)C(Cl)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11414427
|
Thermo Scientific Alfa Aesar
B20454.03 |
1 g |
44.80 EUR
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11424427
|
Thermo Scientific Alfa Aesar
B20454.06 |
5 g | N/A | ||||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
ApplicationsIt is employed as an active pharmaceutical intermediate.
Solubility
Not miscible or difficult to mix in water.
Notes
Moisture sensitive. Store away from alcohols, amines, oxidizing agents and active metals. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
Chemical Identifiers
216144-68-2 | |
226.55 | |
DPDAEMNQJYDHRQ-UHFFFAOYSA-N | |
2734709 | |
FC1=CC(=CC=C1C(F)(F)F)C(Cl)=O |
C8H3ClF4O | |
MFCD00236284 | |
3-fluoro-4-trifluoromethyl benzoyl chloride, 3-fluoro-4-trifluoromethyl benzoylchloride, 3-fluoranyl-4-trifluoromethyl benzoyl chloride, acmc-1cobr, 4-chlorocarbonyl-2-fluorobenzotrifluoride, 3-fluoro-4-trifiuoromethyl benzoyl chloride, 3-fluoro-4-trifluoromethyl benzoyl cloride, benzoylchloride, 3-fluoro-4-trifluoromethyl, benzoyl chloride, 3-fluoro-4-trifluoromethyl | |
3-fluoro-4-(trifluoromethyl)benzoyl chloride |
Specifications
216144-68-2 | |
151°C to 153°C | |
C8H3ClF4O | |
MFCD00236284 | |
Moisture sensitive | |
Not miscible or difficult to mix in water. | |
FC1=CC(=CC=C1C(F)(F)F)C(Cl)=O | |
226.55 | |
226.56 | |
3-Fluoro-4-(trifluoromethyl)benzoyl chloride |
1.486 | |
81°C (177°F) | |
1.4656 | |
UN3265 | |
3-fluoro-4-trifluoromethyl benzoyl chloride, 3-fluoro-4-trifluoromethyl benzoylchloride, 3-fluoranyl-4-trifluoromethyl benzoyl chloride, acmc-1cobr, 4-chlorocarbonyl-2-fluorobenzotrifluoride, 3-fluoro-4-trifiuoromethyl benzoyl chloride, 3-fluoro-4-trifluoromethyl benzoyl cloride, benzoylchloride, 3-fluoro-4-trifluoromethyl, benzoyl chloride, 3-fluoro-4-trifluoromethyl | |
DPDAEMNQJYDHRQ-UHFFFAOYSA-N | |
3-fluoro-4-(trifluoromethyl)benzoyl chloride | |
2734709 | |
97% |
Safety and Handling
GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
P210-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
H227-H314
missing translation for 'dotInformation' : Hazard Class: 8; Packaging Group: III
missing translation for 'einecsNumber' : 000-000-0
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures