Learn More
3-Hydroxy-4-iodobenzoic acid, 97%, Thermo Scientific™
Product Code. 15473697
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
Product Code. 15473697
Brand: Thermo Scientific Alfa Aesar H27502.MD
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 58123-77-6
Molecular Formula C7H5IO3
Molecular Weight (g/mol) 264.02
MDL Number MFCD02068386
InChI Key UABBBWVTEWIIMN-UHFFFAOYSA-N
Synonym 3-hydroxy-4-iodo-benzoic acid, benzoic acid, 3-hydroxy-4-iodo, acmc-1ay71, ksc497o7h, 4-iodo-3-hydroxybenzoic acid, 4-iodo-3-hydroxy-benzoic acid, 3-hydroxy-4-iodobenzoic acid, 4-iodo-3-hydroxy benzoic acid
PubChem CID 736854
IUPAC Name 3-hydroxy-4-iodobenzoic acid
SMILES OC(=O)C1=CC(O)=C(I)C=C1
Specifications
Melting Point 225°C to 231°C
Quantity 250 mg
Sensitivity Light sensitive
Formula Weight 264.02
Percent Purity 97%
Chemical Name or Material 3-Hydroxy-4-iodobenzoic acid