Learn More
3-Methoxybenzoic acid, 99%
Description
3-Methoxybenzoic acid is used in the synthesis and characterization of 3-methoxybenzoates of europium (III) and gadolinium (III)1. It was used in conversion of aromatic carboxylic acids into methyl esters and reduction to the corresponding primary alcohols using a sodium borohydride-THF-methanol system
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
3-Methoxybenzoic acid is used in the synthesis and characterization of 3-methoxybenzoates of europium (III) and gadolinium (III)1. It was used in conversion of aromatic carboxylic acids into methyl esters and reduction to the corresponding primary alcohols using a sodium borohydride-THF-methanol system
Solubility
Soluble in 95% ethanol (50 mg/ml), water (2 mg/ml at 25°C) and methanol.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with bases and oxidizing agents.
Specifications
Specifications
| Melting Point | 104°C to 108°C |
| Boiling Point | 170°C to 172°C (10 mmHg) |
| Odor | Odorless |
| Quantity | 1000 g |
| Beilstein | 508838 |
| Solubility Information | Soluble in 95% ethanol (50mg/ml),water (2mg/ml at 25°C) and methanol. |
| Formula Weight | 152.15 |
| Percent Purity | 99% |
| Chemical Name or Material | 3-Methoxybenzoic acid |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.