missing translation for 'onlineSavingsMsg'
Learn More

3'-Methoxybiphenyl-4-carboxaldehyde, 96%, Thermo Scientific™

Product Code. 15427257
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 209863-09-2
Molecular Formula C14H12O2
Molecular Weight (g/mol) 212.248
MDL Number MFCD03424619
InChI Key LHVLDSOAJZLBMM-UHFFFAOYSA-N
Synonym 4-3-methoxyphenyl benzaldehyde, 3'-methoxy-1,1'-biphenyl-4-carbaldehyde, 3'-methoxybiphenyl-4-carboxaldehyde, 3'-methoxy-biphenyl-4-carbaldehyde, 3'-methoxybiphenyl-4-carbaldehyde, 3'-methoxy-biphenyl-4-carboxaldehyde, 3'-methoxy-1,1'-biphenyl-4-carboxaldehyde, pubchem10290, 3-4-formylphenyl anisole, amtda053
PubChem CID 2759546
IUPAC Name 4-(3-methoxyphenyl)benzaldehyde
SMILES COC1=CC=CC(=C1)C2=CC=C(C=C2)C=O

Specifications

Flash Point >110°C (230°F)
Quantity 1 g
Sensitivity Air Sensitive
Formula Weight 212.25
Percent Purity 96%
Chemical Name or Material 3'-Methoxybiphenyl-4-carboxaldehyde
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'