missing translation for 'onlineSavingsMsg'
Learn More

3-Methoxypropionitrile, 98%, Thermo Scientific Chemicals

27.94 EUR - 331.65 EUR

Chemical Identifiers

CAS 110-67-8
Molecular Formula C4H7NO
Molecular Weight (g/mol) 85.11
MDL Number MFCD00001958
InChI Key OOWFYDWAMOKVSF-UHFFFAOYSA-N
Synonym 3-methoxypropionitrile, propanenitrile, 3-methoxy, 2-cyanoethyl methyl ether, 3-methoxypropylnitrile, beta-methoxypropionitrile, 1-cyano-2-methoxyethane, propionitrile, 3-methoxy, 3-methoxypropannitril, 3-methoxypropiononitrile, beta-methyoxypropionitrile
PubChem CID 61032
IUPAC Name 3-methoxypropanenitrile
SMILES COCCC#N
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10347181
View Documents
Thermo Scientific Acros
257441000
100 mL Glass bottle
27.94 EUR
100mL
Please to purchase this item. Need a web account? Register with us today!
10707592
View Documents
Thermo Scientific Acros
257445000
500 mL Glass bottle
87.33 EUR
500mL
Please to purchase this item. Need a web account? Register with us today!
10197463
View Documents
Thermo Scientific Acros
257440025
2.5 L Glass bottle
331.65 EUR
2.50 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

110-67-8
85.11
OOWFYDWAMOKVSF-UHFFFAOYSA-N
61032
COCCC#N
C4H7NO
MFCD00001958
3-methoxypropionitrile, propanenitrile, 3-methoxy, 2-cyanoethyl methyl ether, 3-methoxypropylnitrile, beta-methoxypropionitrile, 1-cyano-2-methoxyethane, propionitrile, 3-methoxy, 3-methoxypropannitril, 3-methoxypropiononitrile, beta-methyoxypropionitrile
3-methoxypropanenitrile

Specifications

110-67-8
100.0
Colorless to Yellow
164.0°C to 165.0°C
Authentic
C4H7NO
1.4020 to 1.4040
0.938
Solubility in water: 335g/L (20°C)
COCCC#N
85.11
85.11
Liquid
97.5
-62.9°C
0.9380g/mL
66°C
98%
CH3OCH2CH2CN
MFCD00001958
3-methoxypropionitrile, propanenitrile, 3-methoxy, 2-cyanoethyl methyl ether, 3-methoxypropylnitrile, beta-methoxypropionitrile, 1-cyano-2-methoxyethane, propionitrile, 3-methoxy, 3-methoxypropannitril, 3-methoxypropiononitrile, beta-methyoxypropionitrile
OOWFYDWAMOKVSF-UHFFFAOYSA-N
3-methoxypropanenitrile
61032
98%
3-Methoxypropionitrile, 98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear eye protection/face protection.
Wash face,hands and any exposed skin thoroughly after handling.
If eye irritation persists: Get medical advice/attention.

GHS Signal Word: Warning

missing translation for 'einecsNumber' : 203-790-7

missing translation for 'rtecsNumber' : TZ4870000

missing translation for 'tsca' : TSCA

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.