missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-3-methoxybutanol, 99%, Thermo Scientific Chemicals

27.94 EUR - 71.96 EUR

Chemical Identifiers

CAS 56539-66-3
MDL Number MFCD00044771
InChI Key MFKRHJVUCZRDTF-UHFFFAOYSA-N
Synonym 3-methoxy-3-methyl-1-butanol, 3-methyl-3-methoxybutanol, 3-methoxy-3-methylbutanol, 1-butanol, 3-methoxy-3-methyl, methoxymethylbutanol, 3-methoxy-3-methyl-butan-1-ol, acmc-1bbqk, dsstox_cid_24389, dsstox_rid_80190, dsstox_gsid_44389
PubChem CID 62118
IUPAC Name 3-methoxy-3-methylbutan-1-ol
SMILES CC(C)(CCO)OC
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10083220
View Documents
Thermo Scientific Acros
259752500
250 mL Glass bottle
27.94 EUR
250mL
Please to purchase this item. Need a web account? Register with us today!
10006043
View Documents
Thermo Scientific Acros
259750010
1 L Glass bottle
71.96 EUR
1 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

56539-66-3
MFKRHJVUCZRDTF-UHFFFAOYSA-N
62118
CC(C)(CCO)OC
MFCD00044771
3-methoxy-3-methyl-1-butanol, 3-methyl-3-methoxybutanol, 3-methoxy-3-methylbutanol, 1-butanol, 3-methoxy-3-methyl, methoxymethylbutanol, 3-methoxy-3-methyl-butan-1-ol, acmc-1bbqk, dsstox_cid_24389, dsstox_rid_80190, dsstox_gsid_44389
3-methoxy-3-methylbutan-1-ol

Specifications

56539-66-3
100.0
Colorless
174.0°C
Authentic
C6H14O2
1.4260 to 1.4280
0.926
Solubility in water: freely soluble
CC(C)(CCO)OC
118.17
7.35 mPa.s (20°C)
99%
3-Methyl-3-methoxybutanol, 99%
98.5
-50.0°C
0.9260g/mL
71°C
98.5% min. (GC)
(CH3)2C(OCH3)CH2CH2OH
MFCD00044771
3-methoxy-3-methyl-1-butanol, 3-methyl-3-methoxybutanol, 3-methoxy-3-methylbutanol, 1-butanol, 3-methoxy-3-methyl, methoxymethylbutanol, 3-methoxy-3-methyl-butan-1-ol, acmc-1bbqk, dsstox_cid_24389, dsstox_rid_80190, dsstox_gsid_44389
MFKRHJVUCZRDTF-UHFFFAOYSA-N
3-methoxy-3-methylbutan-1-ol
62118
118.17
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye irritation.

GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Wear face protection.

GHS Signal Word: Warning

missing translation for 'einecsNumber' : 260-252-4

missing translation for 'tsca' : TSCA

Product Identifier
  • 3-Methyl-3-methoxybutanol

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2

Hazard Statement
  • H319-Causes serious eye irritation.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only