missing translation for 'onlineSavingsMsg'
Learn More

3-Oxabicyclo[3.1.0]hexane-2,4-dione, 98%, Thermo Scientific Chemicals

Product Code. 10562174
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5617-74-3
Molecular Formula C5H4O3
Molecular Weight (g/mol) 112.09
MDL Number MFCD00126929
InChI Key ZRMYHUFDVLRYPN-UHFFFAOYSA-N
Synonym 3-oxabicyclo 3.1.0 hexane-2,4-dione, 1,2-cyclopropanedicarboxylic anhydride, oxabicyclohexanedione, pubchem18320, cyclopropane-1,2-dicarboxylic anhydride, 2,4-dioxo-3-oxabicyclo 3.1.0 hexane, 3-oxa-bicyclo 3.1.0 hexane-2,4-dione, 3-oxabicyclo 3.1.0-hexane-2,4-dione, 3-oxa-bicyclo 3.1.0-hexane-2,4-dione
PubChem CID 2734721
IUPAC Name 3-oxabicyclo[3.1.0]hexane-2,4-dione
SMILES C1C2C1C(=O)OC2=O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 54°C to 60°C
Color White
Boiling Point 100°C to 102°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 5 g
Solubility Information Solubility in water: insoluble
Formula Weight 112.09
Percent Purity 98%
Physical Form Powder, Crystals or Chunks
Chemical Name or Material 3-Oxabicyclo[3.1.0]hexane-2, 4-dione, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 3-Oxabicyclo¢3.1.0!hexane-2,4-dione
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'