missing translation for 'onlineSavingsMsg'
Learn More

3-Phenoxypropionic acid, 98+%

Product Code. 11463438
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11463438

Brand: Thermo Scientific Alfa Aesar A17842.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7170-38-9 | C9H9O3 | 165.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7170-38-9
Molecular Formula C9H9O3
Molecular Weight (g/mol) 165.17
MDL Number MFCD00002769
InChI Key BUSOTUQRURCMCM-UHFFFAOYSA-M
Synonym 3-phenoxypropionic acid, propanoic acid, 3-phenoxy, beta-phenoxypropionic acid, 3-penoxypropionic acid, 3-phenoxy-propionic acid, 3-phenoxypropionicacid, b-phenoxypropionate, 3-phenoxypropionate, 3-phenoxy-propanoate, beta-phenoxypropionate
PubChem CID 81596
IUPAC Name 3-phenoxypropanoic acid
SMILES [O-]C(=O)CCOC1=CC=CC=C1

Specifications

Melting Point 96°C to 99°C
Boiling Point 243°C to 245°C
Quantity 5 g
Beilstein 1638040
Formula Weight 166.17
Percent Purity ≥98%
Chemical Name or Material 3-Phenoxypropionic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.