Learn More
Sampling Systems™ 304 Stainless Steel Storage Containers without Toggle Clamps
Shop All Sampling Systems Products
Click to view available options
Capacity (Metric):
20 L
25 L
30 L
50 L
75 L
100 L
Unit Size:
Each
Description
- Manufactured from durable 304 grade stainless steel
- Crevice-free interior
- Close fitting recessed lid
- Each with two sturdy handles
- Flat bottom with chime ring to give added protection
- Light duty construction
- Available in a wide range of capacities to fit your specific needs
Specifications
Specifications
| Closure Included | Yes |
| Product Type | Storage container |
| Closure Type | Recessed Lid with Handle |
| Capacity (Metric) | 50 L |
| For Use With (Application) | Multi-purpose Storage |
| Material | Stainless Steel |
| Height (Metric) | 560 mm |
| Diameter (Metric) Outer | 350 mm |