Learn More
Watson-Marlow Pumps 318MC Series Pumphead
318MC Pumphead features high accuracy microcassettes for manifold tubing for flow rates up to 53 mL/min.
Brand:  Watson-Marlow Pumps 033.6854.000
 View more versions of this product
Product Code. 12831921
  • 820.00 EUR / Each
Estimated Shipment: 27-02-2025
Description
  • Designed for multi-channel low-flow applications.
  • For simple tube change without disturbing other channels, cassettes are pre-loaded with tubing, which is available in twenty sizes and three different materials.
  • Extension pumpheads snap fit in seconds, and two MCX (five channel) or three MCX3 (three channel extension pumpheads can be added), depending on the power of the drive.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
36 mL/min.
Multi-channel Pump Head
Glass Filled Polypropylene, IXEF (Polyarylamide), Stainless Steel
318MCX
Electroless Nickel Plated Hardened Steel, Glass Filled Nylon