Learn More
Steris Corporation 36ROLL Autoclave indicator tape lead free green striped 25mm x 50m (w x l) colour change to brown
Brand:  Steris Corporation 0163AB
36ROLL Autoclave indicator tape lead free green striped 25mm x 50m (w x l) colour change to brown
Product Code. 17866277
  • ONLINE EXCLUSIVE PRICE
    230.00 EUR
    199.00 EUR / 36 rolls
    Save 31.00 EUR
    13% Off
Only null left
Estimated Shipment: 05-02-2026