Learn More
3M™ Disposable Protective Coveralls, 4540 Series
With premium dual material technology and offering CE Category III, Type 5/6 protection, the Protective Coverall 4540 Series is ideal for applications where both high levels of protective performance and comfort are desired.
Brand: 3M™ 7000089668
This item is not returnable.
View return policy
Description
- Premium dual material technology provides both protective performance and comfort
- Excellent barrier against dry particles and limited liquid chemical splash (CE Type 5/6)
- Soft and light, super low-linting laminated microporous material
- Back panel made of breathable SMMMS material for improved air circulation and reduced heat stress
- Two-way zipper with fully sealable storm flap
- Three-panel hood design for enhanced compatability with complimentary PPE
- Knitted cuffs and elasticated waist and ankles for increased comfort and freedom of movement
- Anti-static treated
- Low linting