Learn More
Cole-Parmer™ Reference Filling Solution
Shop All Cole Parmer Products
Click to view available options
Quantity:
125 mL
Unit Size:
Each
Description
High-temperature 1.5M KCl gel saturated with AgCl; 125ml
Specifications
Specifications
| Composition | 3M KCl Gel Saturated with AgCl |
| Concentration | 3M KCl gel saturated with AgCl |
| For Use With (Equipment) | Autoclavable Fermentation pH Electrodes |
| Packaging | 1 Each |
| Quantity | 125 mL |