Learn More
3M™ Qualitative Fit-Test Kit Accessories
Click to view available options
For Use With (Application):
Provide secure hood environment
Quantitative Fit Testing
Used to check face-to-respirator seal on any particulate respirator or gas/vapor respirator with a particulate filter
Product Type:
Collar
Fit Test Adapter
Nebulizer
Respirator Test Hood
Unit Size:
Each
Pack of 2
Pack of 3
Description
- Test kits and accessories designed to help your organization meet OSHA fit testing requirements for respirators
- The products can be used to check face-to-respirator seal on any particulate respirator or gas/vapor respirator with a particulate prefilter
- 3M Catalog# 601, FT-13, FT-14, FT-15
Specifications
Specifications
| Product Type | Nebulizer |
| For Use With (Application) | Used to check face-to-respirator seal on any particulate respirator or gas/vapor respirator with a particulate filter |
| For Use With (Equipment) | Qualitative Fit Test Apparatus FT10 and FT30 |
| Model | FT-13 |
| Quantity | 3/Cs. |