Learn More
4-(1-Piperazinyl)aniline, 95%, Thermo Scientific Chemicals
27.10 EUR - 315.57 EUR
Chemical Identifiers
CAS | 67455-41-8 |
---|---|
Molecular Formula | C10H15N3 |
Molecular Weight (g/mol) | 177.25 |
MDL Number | MFCD01365906 |
InChI Key | VAVOYRCCWLRTMS-UHFFFAOYSA-N |
Synonym | 4-piperazin-1-yl aniline, 1-4-aminophenyl piperazine, 4-piperazinoaniline, 4-piperazin-1-yl-phenylamine, 4-1-piperazinyl aniline, benzenamine,4-1-piperazinyl, 4-piperazinylphenylamine, benzenamine, 4-1-piperazinyl, 4-piperazin-1-ylphenylamine, 1-p-aminophenyl piperazine |
PubChem CID | 422925 |
IUPAC Name | 4-piperazin-1-ylaniline |
SMILES | NC1=CC=C(C=C1)N1CCNCC1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15451398
|
Thermo Scientific Alfa Aesar
H55224.MD |
250 mg | N/A | ||||||
15431398
|
Thermo Scientific Alfa Aesar
H55224.03 |
1 g |
78.59 EUR
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15441398
|
Thermo Scientific Alfa Aesar
H55224.06 |
5 g |
315.57 EUR
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
4-Piperazinoaniline(CAS#67455-41-8)is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-Piperazinoaniline(CAS#67455-41-8)is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.
Notes
Reactive with oxidizing agents. Keep at room temperature.
Chemical Identifiers
67455-41-8 | |
177.25 | |
VAVOYRCCWLRTMS-UHFFFAOYSA-N | |
422925 | |
NC1=CC=C(C=C1)N1CCNCC1 |
C10H15N3 | |
MFCD01365906 | |
4-piperazin-1-yl aniline, 1-4-aminophenyl piperazine, 4-piperazinoaniline, 4-piperazin-1-yl-phenylamine, 4-1-piperazinyl aniline, benzenamine,4-1-piperazinyl, 4-piperazinylphenylamine, benzenamine, 4-1-piperazinyl, 4-piperazin-1-ylphenylamine, 1-p-aminophenyl piperazine | |
4-piperazin-1-ylaniline |
Specifications
67455-41-8 | |
Purple to White | |
MFCD01365906 | |
VAVOYRCCWLRTMS-UHFFFAOYSA-N | |
4-piperazin-1-ylaniline | |
422925 | |
97% | |
4-(1-Piperazinyl)aniline |
119°C to 123°C | |
C10H15N3 | |
4-piperazin-1-yl aniline, 1-4-aminophenyl piperazine, 4-piperazinoaniline, 4-piperazin-1-yl-phenylamine, 4-1-piperazinyl aniline, benzenamine,4-1-piperazinyl, 4-piperazinylphenylamine, benzenamine, 4-1-piperazinyl, 4-piperazin-1-ylphenylamine, 1-p-aminophenyl piperazine | |
NC1=CC=C(C=C1)N1CCNCC1 | |
177.25 | |
177.25 | |
Powder |
Safety and Handling
GHS H Statement
H301-H318-H315-H335
Toxic if swallowed.
Causes serious eye damage.
Causes skin irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P362-P501c
H302-H315-H318-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only