Learn More
Memmert™ 4-20 mA Current Loop Interface for UF750plus, Actual Value
Product Code. 17529625 Shop All Memmert Products
Click to view available options
Amperage:
4 to 20 mA
Unit Size:
Each
Product Code. 17529625
Brand: Memmert™ UF750plusV3
Only null left
Add to basket
Add to basket
Temperature Output for external temperature monitoring.
  • Temperature controller actual value
Specifications
For Use With (Equipment) UF750plus Oven
Type Current Loop Interface
Amperage 4 to 20 mA