missing translation for 'onlineSavingsMsg'
Learn More
4-(3-Ethyl-1,2,4-oxadiazol-5-yl)piperidine, 95%, Thermo Scientific™ Product Code.: 11459560

4-(3-Ethyl-1,2,4-oxadiazol-5-yl)piperidine, 95%, Thermo Scientific™

Product Code. 11459560
250 mg
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy

Product Code. 11459560

Brand: Thermo Scientific Alfa Aesar H50874.MD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
4-(3-Ethyl-1,2,4-oxadiazol-5-yl)piperidine is used as organic chemical synthesis intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 912761-48-9
Molecular Formula C9H15N3O
Molecular Weight (g/mol) 181.239
MDL Number MFCD08061039
InChI Key PTQDWAPTOMWGGM-UHFFFAOYSA-N
PubChem CID 24208846
IUPAC Name 3-ethyl-5-piperidin-4-yl-1,2,4-oxadiazole
SMILES CCC1=NOC(=N1)C2CCNCC2

Specifications

Quantity 250 mg
Synonym 4-3-ethyl-1,2,4-oxadiazol-5-yl piperidine, 3-ethyl-5-4-piperidyl-1,2,4-oxadiazole, 3-ethyl-5-piperidin-4-yl-1,2,4-oxadiazole, piperidine, 4-3-ethyl-1,2,4-oxadiazol-5-yl, pubchem18474, 4-3-ethyl-1,2,4 oxadiazol-5-yl-piperidine, piperidine,4-3-ethyl-1,2,4-oxadiazol-5-yl, 4-3-ethyl-1,2,4 oxadiazol-5-yl-piperidine hydrochloride
Solubility Information Slightly soluble in water.
Formula Weight 181.24
Percent Purity 95%
Chemical Name or Material 4-(3-Ethyl-1,2,4-oxadiazol-5-yl)piperidine

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

4 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberE30S035 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number
Lot NumberA26S053 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog Number
Lot NumberJ18Q023 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog Number
Lot NumberJ22T003 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog Number
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4-(3-Ethyl-1,2,4-oxadiazol-5-yl)piperidine, 95%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.