missing translation for 'onlineSavingsMsg'
Learn More

4-(4-Morpholinyl)-2-(trifluoromethyl)aniline, 97%, Thermo Scientific™

Code produit. 15438305
missing translation for 'orderingAttributeHoverText'
Quantity:
250 mg
1 g
5 g
missing translation for 'unitSize'
1g
250mg
5g
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

Code produit. 15438305

missing translation for 'mfr': Thermo Scientific Alfa Aesar H33081.MD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
As pharmaceutical intermediate.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78335-26-9
Molecular Formula C11H13F3N2O
Molecular Weight (g/mol) 246.233
MDL Number MFCD01436021
InChI Key DZEZKCZKDBIQSW-UHFFFAOYSA-N
Synonym 4-morpholin-4-yl-2-trifluoromethyl aniline, 4-morpholin-4-yl-2-trifluoromethyl-phenylamine, 4-morpholino-2-trifluoromethyl aniline, 4-4-morpholinyl-2-trifluoromethyl aniline, benzenamine, 4-4-morpholinyl-2-trifluoromethyl
PubChem CID 822225
IUPAC Name 4-morpholin-4-yl-2-(trifluoromethyl)aniline
SMILES C1COCCN1C2=CC(=C(C=C2)N)C(F)(F)F

Specifications

Quantity 250 mg
Formula Weight 246.23
Percent Purity 97%
Chemical Name or Material 4-(4-Morpholinyl)-2-(trifluoromethyl)aniline
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.