missing translation for 'onlineSavingsMsg'
Learn More
4,5-Bis(di-tert-butylphosphino)-9,9-dimethylxanthene, 99%, Thermo Scientific Chemicals Product Code.: 11317249

4,5-Bis(di-tert-butylphosphino)-9,9-dimethylxanthene, 99%, Thermo Scientific Chemicals

Product Code. 11317249
1 g
Click to view available options
Quantity:
500 mg
1 g
5 g
This item is not returnable. View return policy

Product Code. 11317249

Brand: Thermo Scientific Alfa Aesar H27420.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 856405-77-1
Molecular Formula C31H48OP2
Molecular Weight (g/mol) 498.672
MDL Number MFCD07781995
InChI Key ZEIZANJFJXHMNS-UHFFFAOYSA-N
Synonym 9,9-dimethyl-4,5-bis di-tert-butylphosphino xanthene, 9,9-dimethyl-9h-xanthene-4,5-diyl bis di-tert-butylphosphine, t-bu-xantphos, 4,5-bis di-tert-butylphosphino-9,9-dimethylxanthene, 9,9-dimethyl-4,5-bis di-t-butylphosphino xanthene, di-tert-butyl 5-di-tert-butylphosphanyl-9,9-dimethylxanthen-4-yl phosphane, acmc-20alvv, 4,5-bis di-t-butylphosphino-9,9-dimethylxanthene, 4,5-bis di-tert-butylphosphino-9,9-dimethyl-9h-xanthene
PubChem CID 16218202
IUPAC Name ditert-butyl-(5-ditert-butylphosphanyl-9,9-dimethylxanthen-4-yl)phosphane
SMILES CC1(C2=C(C(=CC=C2)P(C(C)(C)C)C(C)(C)C)OC3=C1C=CC=C3P(C(C)(C)C)C(C)(C)C)C

Specifications

Melting Point 153°C to 157°C
Quantity 1 g
Formula Weight 498.68
Percent Purity 99%
Chemical Name or Material 4,5-Bis(di-tert-butylphosphino)-9,9-dimethylxanthene

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10220907 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH27420.ME,H27420.06,H27420.03
Lot Number10195042 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH27420.ME,H27420.06,H27420.03
Lot Number10199790 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH27420.ME,H27420.06,H27420.03
Lot Number10216065 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH27420.ME,H27420.06,H27420.03
Lot Number10225297 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH27420.ME,H27420.06,H27420.03

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4,5-Bis(di-tert-butylphosphino)-9,9-dimethylxanthene, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.