missing translation for 'onlineSavingsMsg'
Learn More
4'-Aminobenzanilide, 97%, Thermo Scientific Chemicals Product Code.: 11471677

4'-Aminobenzanilide, 97%, Thermo Scientific Chemicals

Product Code. 11471677
5 g
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy

Product Code. 11471677

Brand: Thermo Scientific Alfa Aesar B25663.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

4'-Aminobenzanilide is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff field. It has anticonvulsant property.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4′-Aminobenzanilide is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff field. It has anticonvulsant property.

Solubility
Soluble in DMSO and methanol.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17625-83-1
Molecular Formula C13H12N2O
Molecular Weight (g/mol) 212.252
MDL Number MFCD00035777
InChI Key GTTFJYUWPUKXJH-UHFFFAOYSA-N
Synonym 4'-aminobenzanilide, n-4-aminophenyl benzamide, benzamide, n-4-aminophenyl, 4,4'-diaminobenzoylanilide, n-4-amino-phenyl-benzamide, n-4-aminophenyl-benzamide, 4-aminobenzanilide, 4/'-aminobenzanilide, acmc-209xfr, n-benzoyl 4-aminoanilineShow MoreShow Less
PubChem CID 87196
IUPAC Name N-(4-aminophenyl)benzamide
SMILES C1=CC=C(C=C1)C(=O)NC2=CC=C(C=C2)N

Specifications

Melting Point 127°C to 131°C
Quantity 5 g
Solubility Information Soluble in DMSO and methanol.
Formula Weight 212.25
Percent Purity 97%
Chemical Name or Material 4'-Aminobenzanilide

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

4 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10249080 Certificate TypeCertificate of Analysis Date08/03/2024 Catalog NumberB25663.14,B25663.06
Lot Number10245345 Certificate TypeCertificate of Analysis Date13/01/2023 Catalog NumberB25663.14,B25663.06
Lot Number10236614 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB25663.14,B25663.06
Lot Number10128402 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB25663.14,B25663.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4'-Aminobenzanilide, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.