missing translation for 'onlineSavingsMsg'
Learn More

4-(Benzyloxycarbonylamino)cyclohexanol, 97%, predominantly trans, Thermo Scientific™

Product Code. 11371929
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11371929

Brand: Thermo Scientific Alfa Aesar H26016.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16801-62-0
Molecular Formula C14H19NO3
Molecular Weight (g/mol) 249.31
MDL Number MFCD02728352
InChI Key JNPBKQOVSMBPGE-UHFFFAOYSA-N
Synonym benzyl trans-4-hydroxycyclohexyl carbamate, benzyl 4-hydroxycyclohexyl carbamate, benzyl n-4-hydroxycyclohexyl carbamate, n-cbz-4-amino-cyclohexanol, 4-hydroxy-cyclohexyl-carbamic acid benzyl ester, n-cbz-4-hydroxycyclohexane, 4-hydroxycyclohexyl carbamic acid benzyl ester, benzyl 4-hydroxycyclohexylcarbamate, n-cbz-4-aminocyclohexanol, trans-4-cbz-amino cyclohexan-1-ol
PubChem CID 5175065
IUPAC Name benzyl N-(4-hydroxycyclohexyl)carbamate
SMILES C1CC(CCC1NC(=O)OCC2=CC=CC=C2)O

Specifications

Melting Point 162°C to 164°C
Odor Odorless
Quantity 25 g
Formula Weight 249.31
Percent Purity 97%
Chemical Name or Material 4-(Benzyloxycarbonylamino)cyclohexanol, predominantly trans
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.