missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-3-fluorobenzyl bromide, 96%, Thermo Scientific™

Product Code. 15445538
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 127425-73-4
Molecular Formula C7H5Br2F
Molecular Weight (g/mol) 267.923
MDL Number MFCD11847179
InChI Key GMVNLHLMJSMARX-UHFFFAOYSA-N
Synonym 4-bromo-3-fluorobenzyl bromide, 1-bromo-4-bromomethyl-2-fluorobenzene, 3-fluoro-4-bromobenzyl bromide, 4-bromo-3-fluorobenzylbromide, 4-bromo-1-bromomethyl-3-fluorobenzene, 3-fluoro-4-bromobenzylbromide, 1-bromo-4-bromomethyl-2-fluoro-benzene, benzene, 1-bromo-4-bromomethyl-2-fluoro, ksc493i4n, alpha,4-dibromo-3-fluorotoluene
PubChem CID 11666176
IUPAC Name 1-bromo-4-(bromomethyl)-2-fluorobenzene
SMILES C1=CC(=C(C=C1CBr)F)Br

Specifications

Melting Point 33°C to 36°C
Quantity 1 g
UN Number UN3261
Formula Weight 267.92
Percent Purity 96%
Chemical Name or Material 4-Bromo-3-fluorobenzyl bromide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.