missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-3-methylbenzoic acid, 97%, Thermo Scientific Chemicals

67.50 EUR - 198.00 EUR

Chemical Identifiers

CAS 7697-28-1
Molecular Formula C8H6BrO2
Molecular Weight (g/mol) 214.04
MDL Number MFCD00039526
InChI Key KWVXDZLVCISXIB-UHFFFAOYSA-M
Synonym 4-bromo-m-toluic acid, 4-bromo-3-methyl-benzoic acid, 4-bromo-3-methylbenzoicacid, 3-methyl-4-bromobenzoic acid, 4-bromo-3-methyl benzoic acid, benzoic acid, 4-bromo-3-methyl, pubchem3807, 4bromo-3-methylbenzoic acid, 4-bromo-5-methylbenzoic acid, ksc379g0b
PubChem CID 82131
IUPAC Name 4-bromo-3-methylbenzoic acid
SMILES CC1=CC(=CC=C1Br)C([O-])=O
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10813011
View Documents
Thermo Scientific Acros
434780050
5 g Glass bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10608360
View Documents
Thermo Scientific Acros
434780250
25 g Glass bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7697-28-1
214.04
KWVXDZLVCISXIB-UHFFFAOYSA-M
82131
CC1=CC(=CC=C1Br)C([O-])=O
C8H6BrO2
MFCD00039526
4-bromo-m-toluic acid, 4-bromo-3-methyl-benzoic acid, 4-bromo-3-methylbenzoicacid, 3-methyl-4-bromobenzoic acid, 4-bromo-3-methyl benzoic acid, benzoic acid, 4-bromo-3-methyl, pubchem3807, 4bromo-3-methylbenzoic acid, 4-bromo-5-methylbenzoic acid, ksc379g0b
4-bromo-3-methylbenzoic acid

Specifications

7697-28-1
100.0
Tan to Yellow
96% min. (HPLC)
MFCD00039526
KWVXDZLVCISXIB-UHFFFAOYSA-M
4-bromo-3-methylbenzoic acid
82131
97%
4-Bromo-3-methylbenzoic acid
96.0
215.5°C to 216.4°C
Authentic
C8H6BrO2
4-bromo-m-toluic acid, 4-bromo-3-methyl-benzoic acid, 4-bromo-3-methylbenzoicacid, 3-methyl-4-bromobenzoic acid, 4-bromo-3-methyl benzoic acid, benzoic acid, 4-bromo-3-methyl, pubchem3807, 4bromo-3-methylbenzoic acid, 4-bromo-5-methylbenzoic acid, ksc379g0b
CC1=CC(=CC=C1Br)C([O-])=O
214.04
215.05
Powder
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

missing translation for 'einecsNumber' : 231-713-7

Product Identifier
  • 4-Bromo-3-methylbenzoic acid

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.