missing translation for 'onlineSavingsMsg'
Learn More

4-Bromomethyl-2,6-dichloropyridine, 97%, Thermo Scientific™

Product Code. 10077999 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 10077999

Brand: Thermo Scientific Acros 433440010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 175204-45-2
Molecular Formula C6H4BrCl2N
Molecular Weight (g/mol) 241
MDL Number MFCD00052637
InChI Key XDKGMGJZRFOZRV-UHFFFAOYSA-N
Synonym 4-bromomethyl-2,6-dichloropyridine, pyridine,4-bromomethyl-2,6-dichloro, 4-bromomethyl-2 6-dichloropyridine, 4-bromomethyl-2,6-dichloropyridin, 4-bromomethyl-2,6-dichloro-pyridine, 2,6-dichloro-4-bromomethyl pyridine, 4-bromomethyl-2,6-bis chloranyl pyridine
PubChem CID 2781829
IUPAC Name 4-(bromomethyl)-2,6-dichloropyridine
SMILES C1=C(C=C(N=C1Cl)Cl)CBr

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 42.5°C to 43.2°C
Color Yellow
Boiling Point 118.0°C (0.2 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 241
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 4-Bromomethyl-2, 6-dichloropyridine
Show More Show Less
compliance-icons
Product Identifier
  • 4-Bromomethyl-2,6-dichloropyridine
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.