Learn More
JULABO™ 4-channel EtherNet / RS232 Converter
4-channel EtherNet/RS232 Converter enables connection of up to 4 instruments via RS232 interface cable with connection to an existing network via RJ45 cable
Brand: JULABO™ 8980032
This item is not returnable.
View return policy
Description
- Moxa Nport 4
Specifications
Ethernet, RS232 | |
1 Pk. | |
100 to 240 V AC / 12 V DC |