missing translation for 'onlineSavingsMsg'
Learn More
4-Chloro-2-(trifluoromethyl)benzaldehyde, 97%, Thermo Scientific™ Product Code.: 11471270

4-Chloro-2-(trifluoromethyl)benzaldehyde, 97%, Thermo Scientific™

Product Code. 11471270
1 g
Click to view available options
Quantity:
1 g
10 g
This item is not returnable. View return policy

Product Code. 11471270

Brand: Thermo Scientific Alfa Aesar H26360.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 320-43-4
Molecular Formula C8H4ClF3O
Molecular Weight (g/mol) 208.564
MDL Number MFCD03092987
InChI Key AHFINSWGYAZBOZ-UHFFFAOYSA-N
Synonym 4-chloro-2-trifluoromethyl benzaldehyde, 2-trifluoromethyl-4-chlorobenzaldehyde, benzaldehyde, 4-chloro-2-trifluoromethyl, pubchem2479, acmc-1ad3e, ksc496c8d, 4-chloro-2-trifluoromethyl-benzaldehydeShow MoreShow Less
PubChem CID 2779017
IUPAC Name 4-chloro-2-(trifluoromethyl)benzaldehyde
SMILES C1=CC(=C(C=C1Cl)C(F)(F)F)C=O

Specifications

Refractive Index 1.496
Quantity 1 g
Sensitivity Air Sensitive
Formula Weight 208.57
Percent Purity 97%
Chemical Name or Material 4-Chloro-2-(trifluoromethyl)benzaldehyde

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10144070 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH26360.03,H26360.09
Lot Number10143194 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog NumberH26360.03,H26360.09
Lot Number10144319 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog NumberH26360.03,H26360.09
Lot Number10156667 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog NumberH26360.03,H26360.09
Lot Number10154094 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog NumberH26360.03,H26360.09
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4-Chloro-2-(trifluoromethyl)benzaldehyde, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.