missing translation for 'onlineSavingsMsg'
Learn More
4-Chloro-3-(trifluoromethyl)benzoic acid, 97+%, Thermo Scientific™ Product Code.: 11420058

4-Chloro-3-(trifluoromethyl)benzoic acid, 97+%, Thermo Scientific™

Product Code. 11420058
1 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 11420058

Brand: Thermo Scientific Alfa Aesar B23656.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is used in the synthesis of pyrrolo[2,3-d]pyrimidine derivatives and their antiproliferative activity against melanoma cell line.

Solubility
Slightly soluble in water at 25°C 0.039 g/L.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1737-36-6
Molecular Formula C8H3ClF3O2
Molecular Weight (g/mol) 223.56
MDL Number MFCD01631468
InChI Key PPHHAZOVVZBSCM-UHFFFAOYSA-M
Synonym 4-chloro-3-trifluoromethyl benzoic acid, benzoic acid, 4-chloro-3-trifluoromethyl, 5-carboxy-2-chlorobenzotrifluoride, 4-chloro-3-trifluoromethyl-benzoic acid, 3-trifluoromethyl-4-chlorobenzoic acid, pubchem4588, intermediates-zcf02292, ksc496a8t, 4-chloro-3-trifluoromethyl
PubChem CID 2773857
IUPAC Name 4-chloro-3-(trifluoromethyl)benzoic acid
SMILES [O-]C(=O)C1=CC=C(Cl)C(=C1)C(F)(F)F

Specifications

Melting Point 163°C to 165°C
Quantity 1 g
Solubility Information Slightly soluble in water at 25°C 0.039g/L.
Formula Weight 224.57
Percent Purity ≥97%
Chemical Name or Material 4-Chloro-3-(trifluoromethyl)benzoic acid

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

3 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10141000 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number
Lot Number10166903 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number
Lot NumberC28N29 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4-Chloro-3-(trifluoromethyl)benzoic acid, 97+%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.