missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-5,6-dimethylthieno[2,3-d]pyrimidine, 96%

Produktkode p-7032347
missing translation for 'orderingAttributeHoverText'
Quantity:
250 mg
1 g
5 g
Förpackningsstorlek
1g
250mg
5g
Denne vare kan ikke returneres. Se returpolitik

Produktkode 15428635

missing translation for 'mfr': Thermo Scientific Alfa Aesar H33770.MD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter

Denne vare kan ikke returneres. Se returpolitik

CAS: 108831-68-1 | C8H7ClN2S | 198.67 g/mol

Employed as a intermediate for pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Employed as a intermediate for pharmaceutical.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed containers. With adequate ventilation. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 108831-68-1
Molecular Formula C8H7ClN2S
Molecular Weight (g/mol) 198.67
MDL Number MFCD00464832
InChI Key HYOBKTVPACQCBR-UHFFFAOYSA-N
Synonym 4-chloro-5,6-dimethylthieno 2,3-d pyrimidine, 4-chloro-5,6-dimethyl-thieno 2,3-d pyrimidine, thieno 2,3-d pyrimidine,4-chloro-5,6-dimethyl, thieno 2,3-d pyrimidine, 4-chloro-5,6-dimethyl, 4-chloro-5,6-dimethylthiopheno 2,3-d pyrimidine, acmc-209vig, 4-chloranyl-5,6-dimethyl-thieno 2,3-d pyrimidine
PubChem CID 290212
IUPAC Name 4-chloro-5,6-dimethylthieno[2,3-d]pyrimidine
SMILES CC1=C(C)C2=C(Cl)N=CN=C2S1

Tekniske data

Melting Point 109°C to 110°C
Quantity 250 mg
Solubility Information Slightly soluble in water.
Formula Weight 198.68
Percent Purity 96%
Chemical Name or Material 4-Chloro-5,6-dimethylthieno[2,3-d]pyrimidine

RUO – Research Use Only

Korrektion af produktindhold

Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.

Produkttitel

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.

Mange tak! Din feedback er blevet sendt. Hos Fisher Scientific arbejder vi altid på at forbedre vores indhold for dig. Vi sætter pris på din feedback.