missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Chloro-7-Nitrobenz-2-Oxa-1,3-Diazole, ∽98 to 99%, MP Biomedicals™
Click to view available options
Quantity:
1 g
Specifications
Specifications
CAS | 10199-89-0 |
Assay Percent Range | Approx. 98 to 99% |
Molecular Formula | C6H2ClN3O3 |
Quantity | 1 g |
Synonym | 4-chloro-7-nitrobenzofurazan, nbd chloride, 4-chloro-7-nitrobenzo-2-oxa-1,3-diazole, nbd-cl, 7-chloro-4-nitrobenzofurazan, 2,1,3-benzoxadiazole, 4-chloro-7-nitro, nbd-chloride, nbd-c 1, 1-chloro-4-nitrobenzoxadiazole, 4-nitro-7-chlorobenzofurazan |
InChI Key | IGHBXJSNZCFXNK-UHFFFAOYSA-N |
SMILES | C1=C(C2=NON=C2C(=C1)Cl)[N+](=O)[O-] |
IUPAC Name | 4-chloro-7-nitro-2,1,3-benzoxadiazole |
Molecular Weight (g/mol) | 199.55 |
PubChem CID | 25043 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'