missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-N,N-dicyclohexylbenzamide, 97%, Thermo Scientific™

Product Code. 15426328
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 349089-31-2
Molecular Formula C19H26ClNO
Molecular Weight (g/mol) 319.87
MDL Number MFCD00540918
InChI Key BMQOLGWVHWPNTP-UHFFFAOYSA-N
PubChem CID 2189789
IUPAC Name 4-chloro-N,N-dicyclohexylbenzamide
SMILES ClC1=CC=C(C=C1)C(=O)N(C1CCCCC1)C1CCCCC1

Specifications

Quantity 250 mg
Formula Weight 319.87
Percent Purity 97%
Chemical Name or Material 4-Chloro-N,N-dicyclohexylbenzamide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'