Learn More
4-Chloroaniline hydrochloride, 97%, Thermo Scientific™
Product Code. 11323217
Click to view available options
Quantity:
5 g
25 g
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 20265-96-7
Molecular Formula C6H7Cl2N
Molecular Weight (g/mol) 164.029
MDL Number MFCD00042057
InChI Key ISJBQSJDQZLCSF-UHFFFAOYSA-N
Synonym 4-chloroaniline hydrochloride, p-chloroaniline hydrochloride, 4-chlorobenzenamine hydrochloride, 4-chloroanilinium chloride, benzenamine, 4-chloro-, hydrochloride, p-chloroanilinium chloride, unii-po3w01cfsw, p-chloroaniline hcl, 4-chloroaniline hcl, p-chlorophenylamine hydrochloride
PubChem CID 5284363
IUPAC Name 4-chloroaniline;hydrochloride
SMILES C1=CC(=CC=C1N)Cl.Cl
Specifications
Melting Point >230°C (sublimation)
Odor Characteristic
Quantity 5 g
UN Number UN2018
Beilstein 3560964
Formula Weight 164.03
Percent Purity 97%
Chemical Name or Material 4-Chloroaniline hydrochloride