Learn More
4-Chlorophenylmagnesium bromide, 1M in MeTHF, Thermo Scientific Chemicals
225.00 EUR
Chemical Identifiers
CAS | 873-77-8 |
---|---|
Molecular Formula | C6H4BrClMg |
Molecular Weight (g/mol) | 215.76 |
MDL Number | MFCD00009926 |
InChI Key | CDEMHJCJMMOFMB-UHFFFAOYSA-M |
Synonym | 4-chlorophenylmagnesium bromide, bromo p-chlorophenyl magnesium, bromo 4-chlorophenyl magnesium, p-chlorophenylmagnesium bromide, 4-chlorophenyl magnesium bromide, pubchem24069, 4-chlorophenylmagnesiumbromide, 4-chlorphenylmagnesium bromide, 4-chloro phenylmagnesiumbromide, 4-chlorophenyhnagnesium bromide, grignard reagent |
PubChem CID | 101888 |
IUPAC Name | magnesium;chlorobenzene;bromide |
SMILES | ClC1=CC=C([Mg]Br)C=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15438047
|
Thermo Scientific Alfa Aesar
|
100 mL |
N/A
|
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15448047
|
Thermo Scientific Alfa Aesar
H51163.AP |
500 mL | N/A | ||||||
Description
4-Chlorophenylmagnesium bromide is used as a Grignard reagent in greener solvent, 2-methyltetrahydrofuran (2-MeTHF).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-Chlorophenylmagnesium bromide is used as a Grignard reagent in greener solvent, 2-methyltetrahydrofuran (2-MeTHF).
Solubility
It reacts violently with water.
Notes
Air & moisture sensitive. Incompatible with oxidizing agents.
Chemical Identifiers
873-77-8 | |
215.76 | |
CDEMHJCJMMOFMB-UHFFFAOYSA-M | |
101888 | |
ClC1=CC=C([Mg]Br)C=C1 |
C6H4BrClMg | |
MFCD00009926 | |
4-chlorophenylmagnesium bromide, bromo p-chlorophenyl magnesium, bromo 4-chlorophenyl magnesium, p-chlorophenylmagnesium bromide, 4-chlorophenyl magnesium bromide, pubchem24069, 4-chlorophenylmagnesiumbromide, 4-chlorphenylmagnesium bromide, 4-chloro phenylmagnesiumbromide, 4-chlorophenyhnagnesium bromide, grignard reagent | |
magnesium;chlorobenzene;bromide |
Specifications
873-77-8 | |
MFCD00009926 | |
Air and moisture sensitive | |
It reacts violently with water. | |
ClC1=CC=C([Mg]Br)C=C1 | |
215.76 | |
215.76 | |
4-Chlorophenylmagnesium bromide |
C6H4BrClMg | |
UN2924 | |
4-chlorophenylmagnesium bromide, bromo p-chlorophenyl magnesium, bromo 4-chlorophenyl magnesium, p-chlorophenylmagnesium bromide, 4-chlorophenyl magnesium bromide, pubchem24069, 4-chlorophenylmagnesiumbromide, 4-chlorphenylmagnesium bromide, 4-chloro phenylmagnesiumbromide, 4-chlorophenyhnagnesium bromide, grignard reagent | |
CDEMHJCJMMOFMB-UHFFFAOYSA-M | |
magnesium;chlorobenzene;bromide | |
101888 | |
1M in MeTHF |
Safety and Handling
GHS H Statement
H225-H314-H318
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Causes serious eye damage.
P210-P223-P231+P232-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P335+P334-P363-P370+P378q-P501c
H225-H260-H302-H314-H335
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.