Learn More
4-(Dibenzylamino)benzeneboronic acid, 95%, Thermo Scientific™
29.30 EUR - 79.10 EUR
Chemical Identifiers
CAS | 159191-44-3 |
---|---|
Molecular Formula | C20H20BNO2 |
Molecular Weight (g/mol) | 317.20 |
MDL Number | MFCD06798225 |
InChI Key | KLGQJKOCSXZOBV-UHFFFAOYSA-N |
Synonym | 4-n,n-dibenzylamino phenylboronic acid, 4-dibenzylamino phenyl boronic acid, 4-dibenzylamino phenylboronic acid, 4-dibenzylamino benzeneboronic acid, b-4-bis phenylmethyl amino phenyl-boronic acid, boronic acid, b-4-bis phenylmethyl amino phenyl, acmc-20amuz, n,n-dibenzyl-4-dihydroxyborylaniline, 4-dibenzylamino benzene boronic acid |
PubChem CID | 18979386 |
IUPAC Name | [4-(dibenzylamino)phenyl]boronic acid |
SMILES | OB(O)C1=CC=C(C=C1)N(CC1=CC=CC=C1)CC1=CC=CC=C1 |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
159191-44-3 | |
317.20 | |
KLGQJKOCSXZOBV-UHFFFAOYSA-N | |
18979386 | |
OB(O)C1=CC=C(C=C1)N(CC1=CC=CC=C1)CC1=CC=CC=C1 |
C20H20BNO2 | |
MFCD06798225 | |
4-n,n-dibenzylamino phenylboronic acid, 4-dibenzylamino phenyl boronic acid, 4-dibenzylamino phenylboronic acid, 4-dibenzylamino benzeneboronic acid, b-4-bis phenylmethyl amino phenyl-boronic acid, boronic acid, b-4-bis phenylmethyl amino phenyl, acmc-20amuz, n,n-dibenzyl-4-dihydroxyborylaniline, 4-dibenzylamino benzene boronic acid | |
[4-(dibenzylamino)phenyl]boronic acid |
Specifications
159191-44-3 | |
C20H20BNO2 | |
4-n,n-dibenzylamino phenylboronic acid, 4-dibenzylamino phenyl boronic acid, 4-dibenzylamino phenylboronic acid, 4-dibenzylamino benzeneboronic acid, b-4-bis phenylmethyl amino phenyl-boronic acid, boronic acid, b-4-bis phenylmethyl amino phenyl, acmc-20amuz, n,n-dibenzyl-4-dihydroxyborylaniline, 4-dibenzylamino benzene boronic acid | |
OB(O)C1=CC=C(C=C1)N(CC1=CC=CC=C1)CC1=CC=CC=C1 | |
317.20 | |
317.19 | |
4-(Dibenzylamino)benzeneboronic acid |
184°C to 188°C | |
MFCD06798225 | |
KLGQJKOCSXZOBV-UHFFFAOYSA-N | |
[4-(dibenzylamino)phenyl]boronic acid | |
18979386 | |
95% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Warning