missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-2-methylbenzyl bromide, 97%, Thermo Scientific™

Product Code. p-7029393
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11367009

Brand: Thermo Scientific Alfa Aesar H26849.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
As pharmaceutical Intermediate.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 862539-91-1
Molecular Formula C8H8BrF
Molecular Weight (g/mol) 203.054
MDL Number MFCD03701059
InChI Key DAKPIAWEPVZDNG-UHFFFAOYSA-N
Synonym 1-bromomethyl-4-fluoro-2-methylbenzene, 4-fluoro-2-methylbenzyl bromide, 4-fluoro-2-methylbenzylbromide, benzene, 1-bromomethyl-4-fluoro-2-methyl, 2-methyl-4-fluorobenzyl bromide, 4-fluoro-2-methyl-benzyl bromide, 1-bromomethyl-4-fluoro-2-methyl-benzene, benzene,1-bromomethyl-4-fluoro-2-methyl
PubChem CID 3732707
IUPAC Name 1-(bromomethyl)-4-fluoro-2-methylbenzene
SMILES CC1=C(C=CC(=C1)F)CBr

Specifications

Refractive Index 1.5495
Quantity 1 g
UN Number UN3265
Solubility Information Insoluble in water.
Formula Weight 203.06
Percent Purity 97%
Chemical Name or Material 4-Fluoro-2-methylbenzyl bromide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.