missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-N-methylbenzylamine, 97%

Product Code. 15400688
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 15400688

Brand: Thermo Scientific Alfa Aesar H54369.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 405-66-3 | C8H10FN | 139.173 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 405-66-3
Molecular Formula C8H10FN
Molecular Weight (g/mol) 139.173
MDL Number MFCD03839848
InChI Key SZJIQLSCDIEJFC-UHFFFAOYSA-N
Synonym n-methyl-4-fluorobenzylamine, 4-fluoro-n-methylbenzylamine, 4-fluoro-benzyl-methyl-amine, 1-4-fluorophenyl-n-methylmethanamine, n-4-fluorobenzyl-n-methylamine, 4-fluoro-n-methyl benzylamine, 4-fluorophenyl methyl methyl amine, benzenemethanamine, 4-fluoro-n-methyl, 1-4-fluorophenyl-n-methyl-methanamine, acmc-1aek3
PubChem CID 736860
IUPAC Name 1-(4-fluorophenyl)-N-methylmethanamine
SMILES CNCC1=CC=C(C=C1)F

Specifications

Density 1.056
Boiling Point 180°C to 181°C
Flash Point 71°C (160°F)
Refractive Index 1.499
Quantity 5 g
Sensitivity Air Sensitive
Formula Weight 139.17
Percent Purity 97%
Chemical Name or Material 4-Fluoro-N-methylbenzylamine
compliance-icons compliance-icons
Product Identifier
  • 4-Fluoro-N-methylbenzylamine
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : 4-Fluoro-N-methylbenzylamine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.