missing translation for 'onlineSavingsMsg'
Learn More
4-Hydroxy-2-methylquinoline, 98+%, Thermo Scientific Chemicals Product Code.: 11471998

4-Hydroxy-2-methylquinoline, 98+%, Thermo Scientific Chemicals

Product Code. 11471998
25 g
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy

Product Code. 11471998

Brand: Thermo Scientific Alfa Aesar A14587.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

4-Hydroxy-2-methylquinoline is act as an intermediate in the synthesis of dequalinium chloride and as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Hydroxy-2-methylquinoline is act as an intermediate in the synthesis of dequalinium chloride and as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry conditions in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 607-67-0
Molecular Formula C10H9NO
Molecular Weight (g/mol) 159.19
MDL Number MFCD00006758,MFCD00518775
InChI Key NWINIEGDLHHNLH-UHFFFAOYSA-N
Synonym 2-methylquinolin-4-ol, 4-hydroxy-2-methylquinoline, 2-methyl-4-quinolinol, 4-quinolinol, 2-methyl, 4-hydroxyquinaldine, 2-methyl-4-hydroxyquinoline, 2-methyl-quinolin-4-ol, 2-methylquinolin-4 1h-one, 2-methyl-4 1h-quinolinone, 2-methyl-1,4-dihydroquinolin-4-oneShow MoreShow Less
PubChem CID 69089
SMILES CC1=CC(=O)C2=CC=CC=C2N1

Specifications

Melting Point 234°C to 236°C
Quantity 25 g
Beilstein 118320
Solubility Information Slightly soluble in water.
IUPAC Name 2-methyl-1H-quinolin-4-one
Formula Weight 159.19
Percent Purity ≥98%
Chemical Name or Material 4-Hydroxy-2-methylquinoline

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10239339 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog NumberA14587.22,A14587.06,A14587.14
Lot Number10229693 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA14587.22,A14587.06,A14587.14
Lot Number10172296 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA14587.22,A14587.06,A14587.14
Lot NumberH2600A Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA14587.22,A14587.06,A14587.14
Lot Number10227030 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA14587.22,A14587.06,A14587.14

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4-Hydroxy-2-methylquinoline, 98+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.