Learn More
4-Indolylboronic acid, 95%, Thermo Scientific™
255.00 EUR - 827.00 EUR
Chemical Identifiers
CAS | 220465-43-0 |
---|---|
Molecular Formula | C8H8BNO2 |
Molecular Weight (g/mol) | 160.97 |
InChI Key | USJUQEVUEBCLLR-UHFFFAOYSA-N |
Synonym | indole-4-boronic acid, 4-boronoindole, 1h-indol-4-yl boronic acid, 1h-indole-4-boronic acid, 4-indoleboronic acid, 4-indolylboronic acid, 4-borono-1h-indole, 1h-indol-4-yl-4-boronic acid, boronic acid, 1h-indol-4-yl, b-1h-indol-4-yl-boronic acid |
PubChem CID | 2763173 |
IUPAC Name | 1H-indol-4-ylboronic acid |
SMILES | B(C1=C2C=CNC2=CC=C1)(O)O |
Description
Chemical Identifiers
220465-43-0 | |
160.97 | |
indole-4-boronic acid, 4-boronoindole, 1h-indol-4-yl boronic acid, 1h-indole-4-boronic acid, 4-indoleboronic acid, 4-indolylboronic acid, 4-borono-1h-indole, 1h-indol-4-yl-4-boronic acid, boronic acid, 1h-indol-4-yl, b-1h-indol-4-yl-boronic acid | |
1H-indol-4-ylboronic acid |
C8H8BNO2 | |
USJUQEVUEBCLLR-UHFFFAOYSA-N | |
2763173 | |
B(C1=C2C=CNC2=CC=C1)(O)O |
Specifications
220465-43-0 | |
Authentic | |
Glass Bottle | |
indole-4-boronic acid, 4-boronoindole, 1h-indol-4-yl boronic acid, 1h-indole-4-boronic acid, 4-indoleboronic acid, 4-indolylboronic acid, 4-borono-1h-indole, 1h-indol-4-yl-4-boronic acid, boronic acid, 1h-indol-4-yl, b-1h-indol-4-yl-boronic acid | |
B(C1=C2C=CNC2=CC=C1)(O)O | |
160.97 | |
160.97 | |
4-Indolylboronic acid |
184.0°C to 188.0°C | |
94% min. (HPLC) | |
C8H8BNO2 | |
USJUQEVUEBCLLR-UHFFFAOYSA-N | |
1H-indol-4-ylboronic acid | |
2763173 | |
95% |
Safety and Handling
GHS H Statement Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Call a POISON CENTER or doctor/physician if you feel unwell.
GHS Signal Word: Warning
- 4-Indolylboronic acid, 95%
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.