missing translation for 'onlineSavingsMsg'
Learn More

4-Iodo-2-(trifluoromethyl)benzonitrile, 97%, Thermo Scientific Chemicals

Product Code. 15481928
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

4-Iodo-2-(trifluoromethyl)benzonitrile is used as pharmaceutical intermediates

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Iodo-2-(trifluoromethyl)benzonitrile is used as pharmaceutical intermediates

Solubility
Slightly soluble in water.

Notes
Light Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 101066-87-9
Molecular Formula C8H3F3IN
Molecular Weight (g/mol) 297.019
MDL Number MFCD08458091
InChI Key RBRTXKYFMXVIER-UHFFFAOYSA-N
Synonym 4-iodo-2-trifluoromethyl benzonitrile, 2-cyano-5-iodobenzotrifluoride, benzonitrile, 4-iodo-2-trifluoromethyl, 2-trifluoromethyl-4-iodobenzonitrile, 4-cyano-3-trifluoromethyl-1-iodobenzene, 4-cyano-3-trifluoromethyl iodobenzene, 4-iodo-2-trifluoromethyl benzenecarbonitrile, pubchem4810, acmc-1bqw7, ksc494m4b
PubChem CID 16218337
IUPAC Name 4-iodo-2-(trifluoromethyl)benzonitrile
SMILES C1=CC(=C(C=C1I)C(F)(F)F)C#N

Specifications

Melting Point 50°C to 55°C
Flash Point >110°C (230°F)
Quantity 250 mg
UN Number UN3439
Sensitivity Light sensitive
Solubility Information Slightly soluble in water.
Formula Weight 297.02
Percent Purity 97%
Chemical Name or Material 4-Iodo-2-(trifluoromethyl)benzonitrile

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'