missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Methoxy-2-(trifluoromethyl)benzenesulfonyl chloride, 97%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
4-Methoxy-2-(trifluoromethyl)benzenesulfonyl chloride, is used as a pharmaceutical intermediate.
Solubility
Insoluble in water.
Notes
Moisture Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents, bases.
4-Methoxy-2-(trifluoromethyl)benzenesulfonyl chloride, is used as a pharmaceutical intermediate.
Solubility
Insoluble in water.
Notes
Moisture Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents, bases.
Specifications
Spezifikation
| Melting Point | 57°C to 60°C |
| Quantity | 1 g |
| UN Number | UN3261 |
| Sensitivity | Moisture sensitive |
| Solubility Information | Insoluble in water. |
| Formula Weight | 274.64 |
| Percent Purity | 97% |
| Chemical Name or Material | 4-Methoxy-2-(trifluoromethyl)benzenesulfonyl chloride |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur