missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxy-3-(trifluoromethyl)benzenesulfonyl chloride, 97%, Thermo Scientific™

Product Code. 15488025
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 15488025

Brand: Thermo Scientific Alfa Aesar H32242.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 694514-21-1
Molecular Formula C8H6ClF3O3S
Molecular Weight (g/mol) 274.638
MDL Number MFCD07774167
InChI Key AXNBCYIRTWOTHS-UHFFFAOYSA-N
Synonym 4-methoxy-3-trifluoromethyl benzenesulfonyl chloride, 4-methoxy-3-trifluoromethyl benzenesulphonyl chloride, 4-methoxy-3-trifluoromethyl benzene-1-sulfonyl chloride, benzenesulfonyl chloride, 4-methoxy-3-trifluoromethyl
PubChem CID 22482384
IUPAC Name 4-methoxy-3-(trifluoromethyl)benzenesulfonyl chloride
SMILES COC1=C(C=C(C=C1)S(=O)(=O)Cl)C(F)(F)F

Specifications

Melting Point 52°C to 55°C
Quantity 1 g
UN Number UN3261
Sensitivity Moisture sensitive
Formula Weight 274.64
Percent Purity 97%
Chemical Name or Material 4-Methoxy-3-(trifluoromethyl)benzenesulfonyl chloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.