missing translation for 'onlineSavingsMsg'
Learn More

4-Methylphenyl 2,3,4,6-tetra-O-acetyl-beta-D-thiogalactopyranoside, Thermo Scientific Chemicals

Product Code. 15440497
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 15440497

Brand: Thermo Scientific Alfa Aesar J66025.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 28244-99-7
Molecular Formula C21H26O9S
Molecular Weight (g/mol) 454.49
MDL Number MFCD09750783
InChI Key XRFOIRMXQHXTRL-IFLJBQAJSA-N
PubChem CID 10366693
IUPAC Name [(2R,3S,4S,5R,6S)-3,4,5-triacetyloxy-6-(4-methylphenyl)sulfanyloxan-2-yl]methyl acetate
SMILES CC1=CC=C(C=C1)SC2C(C(C(C(O2)COC(=O)C)OC(=O)C)OC(=O)C)OC(=O)C

Specifications

Melting Point 115°C to 116°C
Color White
Quantity 5 g
Synonym 4-methylphenyl 2,3,4,6-tetra-o-acetyl-b-d-thiogalactopyranoside, 4-methylphenyl 2,3,4,6-tetra-o-acetyl-1-thiohexopyranoside #, 4'-methylphenyl-1-thio-.beta.-d-galactoside, tetraacetate, 4-methylphenyl2,3,4,6-tetra-o-acetyl-b-d-thiogalactopyranoside, 4'-methylphenyl-1-thio-.beta.-d-galactoside, tetraacetate ester, 4-methylphenyl 2,3,4,6-tetra-o-acetyl-1-thio-beta-d-galactopyranoside, p-tolyl 2-o,3-o,4-o,6-o-tetraacetyl-1-thio-beta-d-galactopyranoside, 2r,3s,4s,5r,6s-2-acetoxymethyl-6-p-tolylthio tetrahydro-2h-pyran-3,4,5-triyl triacetate, 2r,3s,4s,5r,6s-3,4,5-tris acetyloxy-6-4-methylphenyl sulfanyl oxan-2-yl methyl acetate
Formula Weight 454.5
Physical Form Powder
Chemical Name or Material 4-Methylphenyl 2,3,4,6-tetra-O-acetyl-beta-D-thiogalactopyranoside

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.